Learn More
CD35 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CD35 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036415
|
Proteintech
30057-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18054505
|
Proteintech
30057-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD35 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD35. This antibody recognizes human antigen. The CD35 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD35 | |
| Western Blot, ELISA | |
| Rabbit | |
| 1378 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Liquid | |
| Human | |
| C3b/C4b receptor, C3BR, CD35, Complement receptor type 1, CR1, KN | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.