missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39/ENTPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 410.97€
Specifications
| Antigen | CD39/ENTPD1 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
19229-1-AP recognizes both the 3 isoforms and soluble form of CD39 (38-478 aa) molecule.
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.Specifications
| CD39/ENTPD1 | |
| Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P49961, P55772, P97687 | |
| 12495, 64519, 953 | |
| CD39/ENTPD1 Fusion Protein Ag6165 | |
| Primary | |
| -20°C | |
| ENTPD1 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ATPDase, CD39, Ecto apyrase, Ecto ATP diphosphohydrolase 1, Ecto ATPase 1, Ecto ATPDase 1, ENTPD1, NTPDase 1 | |
| Entpd1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title