Learn More
CD3e Armenian Hamster, APC-Cy7, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB2-APC7-25ug
This item is not returnable.
View return policy
Specifications
| CD3e | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| Cd3e | |
| Protein A/G Purified | |
| RUO | |
| 12501 | |
| IgG |
| Flow Cytometry | |
| 145-2C11 | |
| APC-Cyanine7 | |
| XP_006510029 | |
| Armenian Hamster | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |