missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Mouse, Clone: PC3/188A, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 916-MSM10-P0
Additional Details : Weight : 0.01000kg
Specifications
CD3e | |
Cell surface and cytoplasmic | |
PC3/188A | |
Unconjugated | |
NP_000724 | |
Mouse | |
20kDa | |
Protein A/G Purified | |
RUO | |
916 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD3E | |
A synthetic peptide corresponding to amino acids 156-168 of the cytoplasmic domain of human CD3-? chain | |
Jurkat cells. Tonsil and lymph node | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |