Learn More
CD4 Mouse anti-Human, Brilliant Violet 750, Clone: SK3 (also known as Leu3a), BD
Mouse Monoclonal Antibody
253.59€ - 410.88€
Specifications
| Antigen | CD4 |
|---|---|
| Clone | SK3 (also known as Leu3a) |
| Concentration | 0.2mg/mL |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 750 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15834979
|
BD Biosciences
566356 |
25 μg |
254.00€ 253.59€ / Each Save 0.41€ 0.16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15824979
|
BD Biosciences
566355 |
50 μg |
412.00€ 410.88€ / Each Save 1.12€ 0.27% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
The antibody was conjugated to BD Horizon BV750 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 750nm. BD Horizon Brilliant BV750 can be excited by the violet laser (405 nm) and detected with a 750/30 nm filter with a 740 nm long pass. Due to spectral differences between labeled cells and beads, using BD™ CompBeads can result in incorrect spillover values when used with BD Horizon BV750 reagents. Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended. BV750Specifications
| CD4 | |
| 0.2mg/mL | |
| Brilliant Violet 750 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Peripheral Blood T Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| SK3 (also known as Leu3a) | |
| Monoclonal | |
| Mouse | |
| Human | |
| L3T4, Leu3a, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55) | |
| IgG1 κ | |
| Affinity Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.