Learn More
CD4 Mouse anti-Human, CoraLite 488, Clone: OKT4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-65134-100TESTS
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral t cells of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specifications
| CD4 | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| OKT4 | |
| CoraLite 488 | |
| 100 Tests | |
| 920 | |
| IgG2b |