missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rabbit anti-Ferret, Clone: 108, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00€ - 541.00€
Specifications
| Antigen | CD4 |
|---|---|
| Clone | 108 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275567
|
Novus Biologicals
NBP2-90819-50UL |
50 μg |
367.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18218977
|
Novus Biologicals
NBP2-90819-100UL |
100 μg |
541.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal specifically detects CD4 in Ferret samples. It is validated for ELISA.Specifications
| CD4 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Ferret | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Recombinant Ferret CD4 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 108 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 920 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title