Learn More
CD40 Mouse, anti-Human, APC, Clone: 5C3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60113AZ.1
Encompass_Preferred
 View more versions of this product
Product Code. 17179121
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • The 5C3 antibody reacts with CD40, a 45 - 50 kDa type I transmembrane glycoprotein that is a member of the tumor necrosis factor receptor (TNFR) superfamily
  • CD40 plays an important role in B cell development and is also involved in the process of B cell selection in germinal centers
  • It facilitates T cell-B cell interactions for co-stimulation and immune regulation through engagement with CD154 (gp39), the CD40 ligand expressed on T cells
  • In the presence of IL-4, or other stimuli, CD40 promotes B cell proliferation, induces immunoglobulin- (Ig) isotype class switching and rescues B cells from surface Ig-induced apoptosis
  • CD40 is expressed on the surface of B cells, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, endothelial cells, basal epithelial cells, CD34+ hematopoietic progenitor cells, and primary carcinomas
  • It is expressed at low levels, or not at all, on plasma cells
  • The 5C3 antibody has been used for the activation of B cells in in vitro functional assays or for the induction of B cell proliferation
Specifications
Specifications
CD40
Monoclonal
APC
25 Tests
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human
B Cells
Immunohistochemistry, Flow Cytometry, Functional Assay
5C3