missing translation for 'onlineSavingsMsg'
Learn More

CD40 Rat anti-Mouse, PE, Clone: 3/23, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553791

 View more versions of this product

Product Code. 15837398

  • ONLINE EXCLUSIVE PRICE
    315.00€
    313.51€ / Each
    Save 1.49€
    0.47% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 3/23 clone monoclonal antibody specifically binds to CD40, a 40-50 kDa glycoprotein expressed on B lymphocytes and other antigen-presenting cells. CD40 has been reported to be transiently expressed on activated CD4+ and CD8+ T cells and in some mouse strains, the 3/23 mAb has been reported to react with 5-10% of T lymphocytes in adult mouse, but not neonatal, spleen. CD40 plays a key role in B-cell growth and differentiation where interactions of CD40 with its ligand, CD154, are involved in the initiation, effector, and memory stages of cell-mediated immune responses. In addition, CD40 has been reported to be involved with the triggering of NK cells and NK-T cells. Ligation of CD40 with the 3/23 antibody has been reported to induce splenic B cells to express the costimulatory molecule CD86 (B7-2). In addition, although the 3/23 antibody by itself is a weak B-cell mitogen, it has been reported to synergize markedly with mitogenic anti-IgM, anti-IgD mAb or IL-4 to promote B-cell proliferation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD40
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse CD40 Recombinant Protein
0.2 mg
Immunology
Mouse
IgG2a κ
Flow Cytometry
3/23
PE
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.