missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD42b Monoclonal Antibody (HIP1), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00€ - 392.00€
Specifications
| Antigen | CD42b |
|---|---|
| Clone | HIP1 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055047
|
Invitrogen™
H051T03B01-A |
100 Tests |
392.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054935
|
Invitrogen™
H051T02B01-A |
25 Tests |
178.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD42b Monoclonal antibody specifically detects CD42b in Human samples. It is validated for Flow CytometrySpecifications
| CD42b | |
| 0.2 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P07359 | |
| 2811 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| HIP1 | |
| Flow Cytometry | |
| NovaFluor Blue 510 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP | |
| IgG1 κ | |
| Affinity chromatography | |
| Gp1ba |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title