missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse anti-Mouse, PE, Clone: A20, BD

Mouse Monoclonal Antibody

81.54€ - 260.35€

Specifications

Antigen CD45.1
Clone A20
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15871419
View Documents
BD Biosciences
561872
25 μg
ONLINE EXCLUSIVE PRICE
82.40€
81.54€ / Each
Save 0.86€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15827388
View Documents
BD Biosciences
553776
0.2 mg
ONLINE EXCLUSIVE PRICE
261.00€
260.35€ / Each
Save 0.65€
0.25% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The A20 monoclonal antibody specifically binds to CD45 (Leukocyte Common Antigen) on all leukocytes of mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, DA). This alloantigen was originally named Ly-5.2, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.2 to Ly-5.1 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb A20 has been reported not to react with leukocytes of most other mouse strains, which express the CD45.2 alloantigen. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C,respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The A20 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.1 alloantigen, to antigens and LPS.

Specifications

Specifications

CD45.1
0.2mg/mL
Monoclonal
Mouse
Mouse
SJL mouse thymocytes and splenocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
A20
Flow Cytometry
PE
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.