missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio CD45 Mouse, PE-Cy7, Clone: HI30, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB46-PE7-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922088

  • 295.00€ / 100 tests

Estimated Shipment: 04-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD45
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
Protein A/G Purified
RUO
5788
IgG1 κ
Flow Cytometry
HI30
PE-Cyanine7
NP_002829
Mouse
100 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions