missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio CD45RA Mouse, APC, Clone: HI100, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB45-APC-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932058

  • 141.00€ / 25 tests

Estimated Shipment: 02-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD45RA
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
Protein A/G Purified
RUO
5788
IgG2b κ
Flow Cytometry
HI100
APC
NP_002829
Mouse
25 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions