missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Rabbit anti-Human, Clone: CD47/6362R, Novus Biologicals™
Rabbit Monoclonal Antibody
524.00€
Specifications
| Antigen | CD47 |
|---|---|
| Clone | CD47/6362R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
CD47 Monoclonal specifically detects CD47 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| CD47 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 961 | |
| IgG κ | |
| Protein A or G purified |
| CD47/6362R | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| A recombinant fragment (around aa 18-135) of human CD47 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title