missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48/SLAMF2 Antibody (156-4H9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
252.00€ - 451.00€
Specifications
| Antigen | CD48/SLAMF2 |
|---|---|
| Clone | 156-4H9 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246233
|
Novus Biologicals
NBP1-43403-0.025MG |
0.025 mg |
252.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18245585
|
Novus Biologicals
NBP1-43403-0.1MG |
0.1 mg |
451.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.Specifications
| CD48/SLAMF2 | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Human | |
| BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| CD48 | |
| IgG1 | |
| Affinity Purified |
| 156-4H9 | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 962 | |
| The immunogen for this antibody was CD48. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title