missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CD5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rC5/6976 |
| Conjugate | DyLight 550 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 |
| Host Species | Mouse |
| Immunogen | Recombinant human protein fragment corresponding to CD5 extracellular domain |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?