Learn More
CD5 Mouse anti-Human, Unconjugated, Clone: 4D12C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67278-1-IG-20UL
Description
The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| CD5 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 4D12C10 | |
| Unconjugated | |
| 20 μL | |
| 921 | |
| IgG2b |