missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio CD5 Mouse, APC, Clone: UCHT2, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB15-APC-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15921548

  • 128.00€ / 25 tests

Estimated Shipment: 02-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD5
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD5
Protein A/G Purified
RUO
921
IgG1 κ
Flow Cytometry
UCHT2
APC
NP_055022
Mouse
25 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions