missing translation for 'onlineSavingsMsg'
Learn More

CD51 (Integrin αV) Mouse, anti-Human, FITC, Clone: NKI-M9, STEMCELL Technologies

Mouse Monoclonal Antibody

175.50€ - 438.75€

Specifications

Antigen CD51 (Integrin αV)
Clone NKI-M9
Applications Immunoprecipitation, ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17188741
Encompass_Preferred
View Documents
Stemcell Technologies
60043FI.1
25 Tests
175.50€
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17178741
Encompass_Preferred
View Documents
Stemcell Technologies
60043FI
100 Tests
438.75€
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The NKI-M9 antibody reacts with CD51 (vitronectin receptor α chain, or integrin αV), a type I transmembrane glycoprotein containing two subunits (125 kDa and 24 kDa) which are generated by post-translational cleavage and are linked by a disulfide bond
  • CD51 associates non-covalently with integrin β1 (CD29), β3 (CD61), β5, β6, or β8 to form heterodimeric cell adhesion receptors for extracellular matrix components such as fibrinogen, collagen, fibronectin, laminin, osteopontin, thrombospondin, vitronectin, and von Willebrand factor
  • For example, association of CD51 and CD61 forms the integrin αv/β3 receptor primarily involved in binding vitronectin
  • CD51 is expressed broadly on many types of cells, including endothelial cells, fibroblasts, monocytes, macrophages, platelets (at relatively low levels), and osteoclasts
  • It is also found on hepatoma, melanoma, and neuroblastoma cells
Specifications

Specifications

CD51 (Integrin αV)
Immunoprecipitation, ELISA, Flow Cytometry, Functional Assay
FITC
Human
Primary
NKI-M9
Monoclonal
Mouse
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.