missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD56 Recombinant Rabbit Monoclonal Antibody (201), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
109.00€ - 218.00€
Specifications
| Antigen | CD56 |
|---|---|
| Clone | 201 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019530
![]() |
Invitrogen™
MA552230 |
100 Tests |
218.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019300
|
Invitrogen™
MA552231 |
25 Tests |
109.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD56 Recombinant Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow CytometrySpecifications
| CD56 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Recombinant Human NCAM1/CD56 Protein (Leu 20-Pro603). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ncam1 |
| 201 | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Human | |
| P13591 | |
| 4684 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title