missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITLN2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148999
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
May play a role in the defense system against pathogens.
Specifications
| ITLN2 | |
| Polyclonal | |
| Unconjugated | |
| Endothelial lectin HL-2; HL2; HL-2; intelectin 2; intelectin-2; ITLN2; UNQ2789/PRO7179 | |
| E. coli-derived recombinant mouse Intelectin-2, Ala20-Ser298. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction