Learn More
EnquireBio CD62L / L-Selectin Rat, FITC, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB49-F-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD62L / L-Selectin | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
Sell | |
Protein A/G Purified | |
RUO | |
20343 | |
IgG2a κ |
Flow Cytometry | |
MEL-14 | |
FITC | |
NP_001157531 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |