Learn More
CD62P (P-Selectin) Monoclonal Antibody (AK-4), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-0628-95
Description
Description: The AK-4 monoclonal antibody reacts with human CD62P, a 140 kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils. Applications Reported: The AK-4 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. AK-4 has also been reported in blocking adhesion of platelets in in vitro assays. Applications Tested: This AK-4 antibody has been tested by flow cytometric analysis of resting and thrombin-activated human platelets. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD62P (P-Selectin) Monoclonal antibody specifically detects CD62P (P-Selectin) in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Neutralization
Specifications
| CD62P (P-Selectin) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P16109 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Neutralization | |
| AK-4 | |
| Unconjugated | |
| SELP | |
| CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp | |
| Affinity chromatography | |
| RUO | |
| 6403 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.