Learn More
DIRAS2 Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809401S
Description
DIRAS2 Monoclonal antibody specifically detects DIRAS2 in Human samples. It is validated for Western BlotSpecifications
| DIRAS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96HU8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54769 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI10A6 | |
| Unconjugated | |
| DIRAS2 | |
| 2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2 | |
| Human recombinant protein fragment corresponding to amino acids 1-196 of human DIRAS2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.