Learn More
CD68 Mouse, anti-Human, CoraLite Plus 750, Clone: 3A9A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL750-66231-100UL
Description
The CD68 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD68. This antibody recognizes human, mouse antigen. The CD68 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.
Specifications
| CD68 | |
| 3A9A7 | |
| CoraLite Plus 750 | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| CD68, CD68 molecule, DKFZp686M18236, GP110, Macrosialin, SCARD1 | |
| 100 μL | |
| 968 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |