Learn More
CD68 Mouse, anti-Human, CoraLite Plus 750, Clone: 3A9A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL750-66231-100UL
 View more versions of this product
Product Code. 18048075
  • 450.00€ / 100µL
Description
The CD68 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD68. This antibody recognizes human, mouse antigen. The CD68 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.
Specifications
Specifications
CD68
3A9A7
CoraLite Plus 750
Mouse
Primary
Human, Mouse
155 kDa
IgG1
Western Blot
1000 μg/mL
CD68, CD68 molecule, DKFZp686M18236, GP110, Macrosialin, SCARD1
100 μL
968
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.