Learn More
CD68 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30929-1-AP-20UL
Description
The CD68 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse CD68. This antibody recognizes mouse antigen. The CD68 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| CD68 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| Cd68, CD68 antigen, gp110, Scard1 | |
| 20 μL | |
| 12514 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |