Learn More
CD68 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30929-1-AP-20UL
 View more versions of this product
Product Code. 18057905
  • 145.00€ / 20µL
Description
The CD68 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse CD68. This antibody recognizes mouse antigen. The CD68 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
CD68
500 μg/mL
Rabbit
Primary
Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
Cd68, CD68 antigen, gp110, Scard1
20 μL
12514
Store at -20°C. Stable for one year after shipment.