missing translation for 'onlineSavingsMsg'
Learn More

CD69 Armenian Hamster, FITC, Clone: H1.2F3, enQuireBio™

Armenian Hamster Monoclonal Antibody

131.25€ - 183.75€

Specifications

Antigen CD69
Clone H1.2F3
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15942628
View Documents
EnquireBio
QAB90-F-25ug
25 μg
131.25€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15932628
View Documents
EnquireBio
QAB90-F-100ug
100 μg
183.75€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD69
0.5mg/mL
Monoclonal
Armenian Hamster
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
12515
IgG
Protein A/G Purified
H1.2F3
Flow Cytometry
FITC
RUO
NP_001028294
Cd69
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.