missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Monoclonal Antibody (5-329), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
166.00€ - 411.00€
Specifications
| Antigen | CD74 |
|---|---|
| Clone | 5-329 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16321204
|
Affymetrix eBioscience
62-0748-41 |
25 Tests | |||||||
16331204
![]() |
Invitrogen™
62-0748-42 |
100 Tests |
411.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.Specifications
| CD74 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 972 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 5-329 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Mouse | |
| Human | |
| P04233 | |
| CD74 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| CD74 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title