missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody

Product Code. 12613757 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Unit Size:
100µL
12mL
1mL
500µL
7mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12613757 500 μL Concentrated 500µL
12653757 100 μL Concentrated 100µL
12633757 1 mL Concentrated 1mL
12693747 7 mL Prediluted 7mL
12623757 12 mL Ready-to-Use 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12613757 Brand Epredia™ Brand No. RM9118S1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody.

A disulphide-linked heterodimer, consisting of CD79a/mb-1 and CD79b/B29 polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas.

Host Species: Rabbit

Clone: SP18

Species Reactivity: Human. Does not react with mouse, rat, canine, monkey and opossum

Epitope: N-terminus

Immunogen: Synthetic peptide derived from N-terminal region of human CD79a protein

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a/mb-1 (B-Cell Marker)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP18
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide derived from N-terminal region of human CD79a protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.