missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Mouse anti-Bovine, Goat, Sheep, Clone: CBB/1595, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07571-100ug
This item is not returnable.
View return policy
Description
CD8 beta Monoclonal specifically detects CD8 beta in Bovine, Goat, Sheep samples. It is validated for ELISA.
Specifications
| CD8 beta | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
| Antibody was raised against bovine CD8 beta. | |
| 100 μg | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| 926 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA | |
| CBB/1595 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Goat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction