missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , APC, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
65.48€ - 102.00€
Specifications
| Antigen | CD8 |
|---|---|
| Clone | HIT8a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17257654
|
BD Biosciences
566853 |
25 Tests |
66.35€ 65.48€ / 25 tests Save 0.87€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17247654
|
BD Biosciences
566852 |
100 Tests |
102.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow CytometrySpecifications
| CD8 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |
| HIT8a | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title