missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse, FITC, Clone: SK1, enQuireBio™

Mouse Monoclonal Antibody

248.85€ - 1989.75€

Specifications

Antigen CD8
Clone SK1
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15911598
View Documents
EnquireBio
QAB18-F-25Tests
25 Tests
248.85€
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15901598
View Documents
EnquireBio
QAB18-F-100Tests
100 Tests
472.50€
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15921598
View Documents
EnquireBio
QAB18-F-500Tests
500 Tests
1989.75€
500 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD8
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
925
IgG1 κ
Protein A/G Purified
SK1
Flow Cytometry
FITC
RUO
NP_741969
CD8A
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.