missing translation for 'onlineSavingsMsg'
Learn More

CD80 (B7-1) Monoclonal Antibody (16-10A1), Functional Grade, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

8960.00€ - 14910.00€

Specifications

Antigen CD80 (B7-1)
Clone 16-10A1
Concentration 1 mg/mL
Applications Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17131082
View Documents
Invitrogen
16-0801-025
25 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17141082
View Documents
Invitrogen
16-0801-050
50 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

The 16-10A1 monoclonal antibody reacts with mouse CD80 (B7-1), a 55 kDa member of the Ig superfamily. CD80 is expressed by macrophages, dendritic cells and activated B cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively. Applications Reported: The 16-10A1 antibody has been reported for use in flow cytometric analysis. 16-10A1 has also been reported in blocking of CD80 to its ligands. Applications Tested: The 16-10A1 antibody has been tested by flow cytometric analysis of activated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD80 (B7-1) Monoclonal antibody specifically detects CD80 (B7-1) in Canine, Mouse, Porcine samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Specifications

Specifications

CD80 (B7-1)
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1
IgG
Affinity chromatography
Antibody
16-10A1
Flow Cytometry, Functional Assay, Neutralization
Functional Grade
Armenian Hamster
Canine, Mouse, Pig
Q00609
12519, 397161, 403765
Primary
4° C
CD80
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.