Learn More
Invitrogen™ CD82 Recombinant Rabbit Monoclonal Antibody (072)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530191
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CD82/KAI-1.
This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| CD82 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P27701 | |
| CD82 | |
| Recombinant Human CD82/KAI-1 Protein (Gly111-Leu228). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 072 | |
| Unconjugated | |
| CD82 | |
| 4F9; AA682076; AL023070; C33; C33 antigen; Cd82; CD82 antigen; CD82 antigen (R2 leukocyte antigen antigen detected by monoclonal and antibody IA4)); CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); CD82 molecule; GR15; IA4; Inducible membrane protein R2; KAI1; Kangai 1 (suppression of tumerigenicity 6 prostate) CD82 antigen; kangai 1 (suppression of tumorigenicity 6), prostate; kangai 1 (suppression of tumorigenicity 6, prostate); Kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); metastasis suppressor Kangai-1; Metastasis suppressor Kangai-1 homolog; R2; SAR2; ST6; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan27; tspan-27 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.