Learn More
Invitrogen™ CD86 (B7-2) Monoclonal Antibody (GL1), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
185.00€ - 375.00€
Specifications
| Antigen | CD86 (B7-2) |
|---|---|
| Clone | GL1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17856980
|
Invitrogen™
363-0862-80 |
25 μg |
185.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17866980
![]() |
Invitrogen™
363-0862-82 |
100 μg |
375.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GL1 mAb reacts with mouse CD86, an ∽80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low level on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. CD86 is also expressed by activated mouse T cells and thioglycolate-elicited peritoneal cells. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). This interaction plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation, supports its major contribution during the primary phase of an immune response. Applications Reported: This GL1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This GL1 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. BUV395 is a dye that emits at 395nm and is intended for use on cytometers equipped with an Ultraviolet (355nm) laser. Excitation: 350nm; Emission: 395nm; Laser: Ultraviolet Laser.
CD86 is one of two ligands (the other CD80) for CTLA4 and CD28. CD86 acts as costimulatory molecule in eliciting T-cell help during antigen presentation. Antigen presentation in the absence of sufficient co-stimulation involving CD86/CD80 can induce tolerance. CD80 appears to play a role distinct from CD80 in T helper cell differentiation. CD86 is a type I membrane protein that is a member of the immunoglobulin superfamily. The CD86 protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of CD86 with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of CD86 with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms of CD86. Additional transcript variants have been described for CD86, but their full-length sequences have not been determined. Diseases associated with CD86 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
| CD86 (B7-2) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
| CD86 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD86 |
| GL1 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 395 | |
| Rat | |
| Mouse | |
| P42082 | |
| 12524 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.