missing translation for 'onlineSavingsMsg'
Learn More

CD86 Rat, FITC, Clone: GL-1 (GL1), enQuireBio™

Rat Monoclonal Antibody

116.55€ - 341.25€

Specifications

Antigen CD86
Clone GL-1 (GL1)
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15932158
View Documents
EnquireBio
QAB51-F-25ug
25 μg
116.55€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15922158
View Documents
EnquireBio
QAB51-F-100ug
100 μg
236.25€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15942158
View Documents
EnquireBio
QAB51-F-500ug
500 μg
341.25€
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD86
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
12524
IgG2a κ
Protein A/G Purified
GL-1 (GL1)
Flow Cytometry
FITC
RUO
NP_062261
Cd86
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.