missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8b Monoclonal Antibody (SIDI8BEE), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 62-5273-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD8b Monoclonal antibody specifically detects CD8b in Human, Non-human Primate samples. It is validated for Flow Cytometry
Specifications
| CD8b | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P10966 | |
| Cd8b | |
| Affinity chromatography | |
| RUO | |
| 926 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| SIDI8BEE | |
| Super Bright 436 | |
| Cd8b | |
| BOS_11635; CD8; CD8 antigen 37 kDa chain; CD8a; CD8alpha; Cd8b; CD8B1; CD8beta; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; LYT3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human, Non-human Primate | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction