Learn More
Invitrogen™ MTHFR Recombinant Rabbit Monoclonal Antibody (6Q5C1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558125
Description
Immunogen sequence: RPIFWASRPK SYIYRTQEWD EFPNGRWGNS SSPAFGELKD YYLFYLKSKS PKEELLKMWG EELTSEESVF EVFVLYLSGE PNRNGHKVTC LPWNDEPLAA ETSLLKEELL RVNRQGILTI NSQPNINGKP SSDPIVGWGP SGGYVFQKAY LEFFTSRETA EALLQVLKKY ELRVNYHLVN VKGENITNAP ELQPNAVTWG IFPGREIIQP TVVDPVSFMF WKDEAFALWI ERWGKLYEEE SPSRTIIQYI HDNYFLVNLV DNDFPLDNCL WQVVEDTLEL LNRPTQNARE TEAP.
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
Specifications
| MTHFR | |
| Recombinant Monoclonal | |
| 1.31 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P42898, Q9WU20 | |
| MTHFR | |
| Recombinant protein (or fragment). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6Q5C1 | |
| Unconjugated | |
| MTHFR | |
| 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17769, 362657, 4524 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.