Learn More
CD9 Mouse anti-Human, BUV737, Clone: M-L13, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 749013
Product Code. 17290585
  • 540.00€ / 50µg
Description
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD9
Monoclonal
Brilliant Ultraviolet 737
CD9 antigen (p24); 5H9; BA2; BTCC-1; DRAP-27; GIG2; MIC3; MRP-1; TSPAN29
Human C-ALL Cells
RUO
Human
IgG1 κ
Flow Cytometry
M-L13
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody.