missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Rabbit anti-Human, Mouse, Clone: 10N2L6, Novus Biologicals™
Rabbit Monoclonal Antibody
207.00€ - 511.00€
Specifications
| Antigen | CD9 |
|---|---|
| Clone | 10N2L6 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058413
|
Novus Biologicals
NBP3-15593-20UL |
20 μg |
207.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078345
|
Novus Biologicals
NBP3-15593-100UL |
100 μg |
511.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| CD9 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| A synthetic peptide corresponding to a sequence within amino acids 129-228 of human CD9 (P21926). YNKLKTKDEPQRETLKAIHYALNCCGLAGGVEQFISDICPKKDVLETFTVKSCPDAIKEVFDNKFHIIGAVGIGIAVVMIFGMIFSMILCCAIRRNREMV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10N2L6 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 928 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title