missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (778053), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7335H
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 778053 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD90/Thy1, Gln20-Lys130, Accession # P01831 | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction