missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD96 Recombinant Rabbit Monoclonal Antibody (121)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560315
This item is not returnable.
View return policy
Description
CD96 Recombinant Monoclonal Antibody for ELISA
CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
Specifications
| CD96 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P40200 | |
| Cd96 | |
| Recombinant Human CD96 protein (Met1-Met503). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 121 | |
| Unconjugated | |
| Cd96 | |
| 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10225 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction