missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD97 Recombinant Rabbit Monoclonal Antibody (23GB3740), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

207.00€ - 562.00€

Specifications

Antigen CD97
Clone 23GB3740
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089025
GREENER_CHOICE
View Documents
Invitrogen™
MA553468
100 μL
562.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089337
View Documents
Invitrogen™
MA553469
20 μL
207.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD97 Recombinant Monoclonal antibody specifically detects CD97 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

CD97
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
P48960
976
IgG
Affinity chromatography
Antibody
23GB3740
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1
A synthetic peptide of human CD97 (701-759AA).
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.