Learn More
Invitrogen™ Human CDK13/cyclin K, GST Tag, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK13/cyclin K recombinant human protein (CDK13: 656-1050, cyclin K 1-300) was expressed in insect cells. CDK13/cyclin K recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for CDK13 and cyclin K are NP_003709.3 and NP_001092872.1.
Specifications
Specifications
| Accession Number | O75909, Q14004 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM HEPES with 15mM glutathione, 5mM DTT, 100mM NaCl, 20% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 8621, 8812 |
| Molecular Weight (g/mol) | 73.8-63.0 kDa |
| Name | Human CDK13/cyclin K, GST Tag, His Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.