missing translation for 'onlineSavingsMsg'
Learn More

Cdk7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

386.00€ - 529.00€

Specifications

Antigen Cdk7
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18474061
View Documents
Novus Biologicals
NBP1-86666-25ul
25 μL
386.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18265937
View Documents
Novus Biologicals
NBP1-86666
0.1 mL
529.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cdk7 Polyclonal specifically detects Cdk7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Cdk7
Polyclonal
Rabbit
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
1022
This antibody was developed against Recombinant Protein corresponding to amino acids:GCILAELLLRVPFLPGDSDLDQLTRIFETLGTPTEEQWPDMCSLPDYVTFKSFPGIPLHHIFSAAGDDLLDLIQGLFLFNPCARITATQALKMKYFSNRPGPTPGCQLPRPNCPVETLKEQSNPALAIKRKRTEALEQGGLPKKLI
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500
Unconjugated
RUO
Human
CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit
CDK7
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.