missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21655-1-AP-20UL
This item is not returnable.
View return policy
Description
Could play a role in the terminal differentiation of the intestine.Specifications
| CDX1 | |
| Polyclonal | |
| Unconjugated | |
| CDX1 | |
| caudal type homeobox 1, Caudal type homeobox protein 1, CDX1, Homeobox protein CDX 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1044, 12590 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18111, P47902 | |
| CDX1 | |
| CDX1 Fusion Protein Ag16376 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction