Learn More
CDX2 Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802665S
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Western BlotSpecifications
| CDX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35900 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI5A1 | |
| Unconjugated | |
| KRT20 | |
| caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor | |
| Full length human recombinant protein of human KRT20 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.