missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDY2B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 463.00€
Specifications
| Antigen | CDY2B |
|---|---|
| Dilution | Western Blot 1:1000-1:3000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680348
|
Novus Biologicals
NBP3-05603-100ul |
100 μg |
463.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680598
|
Novus Biologicals
NBP3-05603-20ul |
20 μg |
188.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDY2B Polyclonal antibody specifically detects CDY2B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CDY2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CDY, chromodomain protein, Y chromosome, 2 related, chromodomain protein, Y-linked, 2B, testis-specific chromodomain protein Y 2, testis-specific chromodomain protein Y protein 2 related | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CDY2B (NP_001001722.1). MASQEFEVEAIVDKRQDKNGNTQYLVRWKGYDKQDDTWEPEQHLMNCEKCVHDFNRRQTEKQKKLTWTTTSRIFSNNARRRTSRSTKANYSKNSPKTPVTDKHHRSKNCKLFAASKNVRRKAASTLSDTKNMEIINSTIETLAPDSPFDHKKTVSGFQKLEKLDPIAADQQDTVVFKVTEGKLLRDPLSHPGAEQTGIQNKTQMHPLMSQMSGSVTASMATGSATRKGIVVLIDPLAANGTTDMHTSVPRVKGGQRNITDDSRGQPFIKKMHFTIRLTES | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 203611 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title