Learn More
Vileda Professional™ CE UltraSpeed Pro™ Double Bucket System
Product Code. 17882486
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17882486 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17882486 Supplier Vileda Professional™ Supplier No. 157775
  • ONLINE EXCLUSIVE PRICE
    375.00€
    286.15€ / Pack of 1
    Save 88.85€
    23% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The most effective and easy to use flat mop bucket and wringer system for use on floors, walls and ceilings within Controlled Environments!
Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.
  • Double bucket systems includes two buckets with flat mop wringer and chassis
  • Lightweight double bucket system allows for separation of dirty liquid and clean liquid minimizing cross-contamination
  • Touchless wringing of mops using the patented, fast and efficient wringer/press
  • Small and compact chassis (only 2.29 sq ft) means minimal storage space needed
  • Changeable color-coding clips within the buckets (red, blue, yellow, green)
  • Completely autoclavable with all stainless steel parts
  • Highly durable and withstands all common disinfectants