Learn More
CEA (CEACAM5) Monoclonal Antibody (OTI2B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803181S
Description
CEA (CEACAM5) Monoclonal antibody specifically detects CEA (CEACAM5) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CEA (CEACAM5) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHR5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29115 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B12 | |
| Unconjugated | |
| Sap30bp | |
| 1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; Meconium antigen 100; OTTHUMP00000199033; pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e | |
| Human recombinant protein fragment corresponding to amino acids 1-260 of human SAP30BP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.